Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

905771

Sigma-Aldrich

Dimethylsulfoxonium-3-(methyl)benzoylmethylide

≥95%

Synonym(s):

2-(Dimethyl(oxo)-sulfaneylidene)-1-(m-tolyl)ethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2S
CAS Number:
Molecular Weight:
210.29
UNSPSC Code:
12352101

Assay

≥95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

120-121 °C

functional group

ketone
sulfoxide

storage temp.

−20°C

SMILES string

O=C(C1=CC=CC(C)=C1)C=S(C)(C)=O

General description

Sulfoxonium ylides are highly stable carbene precursors, serving as a safer alternative to diazo compounds as they that do not generate gases as byproducts. Sulfoxonium ylides have been shown to be effective in a wide array of transition metal catalyzed C-H functionalization and coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim Søholm Halskov et al.
Organic letters, 20(8), 2464-2467 (2018-03-28)
Azolopyrimidines are efficiently prepared by direct imidoyl C-H bond activation. Annulations of N-azolo imines with sulfoxonium ylides and diazoketones under redox-neutral conditions and alkynes under oxidizing conditions provide products with various arrangements of nitrogen atoms and carbon substituents. We have
Shuying Ji et al.
Organic letters, 20(18), 5981-5984 (2018-09-13)
A Cp*Co(III)-catalyzed C-H bond functionalization of a range of arenes by employing sulfoxonium ylides as carbene precursors instead of diazo compounds and other carbene precursors has been established. This reaction is highly efficient without any additive, possesses high step and
Panjie Hu et al.
Organic letters, 20(8), 2160-2163 (2018-04-03)
Rh(III)-catalyzed cascade C-H activation of benzoylacetonitriles and annulation with sulfoxonium ylides was realized, leading to selective synthesis of naphthols, 2,3-dihydronaphtho[1,8- bc]pyrans, and naphtho[1,8- bc]pyrans. This step-economic reaction proceeded efficiently under mild and redox-neutral conditions via multiple C-H activations.

Related Content

Ketosulfoxonium ylides serve as versatile carbene equivalents for large-scale reactions, releasing dimethyl sulfoxide biproducts.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service