Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

905534

Sigma-Aldrich

(2-(Phenoxycarbonyl)phenyl)boronic acid

greener alternative

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BO4
CAS Number:
Molecular Weight:
242.04
MDL number:
UNSPSC Code:
12352106

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

150-152 °C

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

(2-(Phenoxycarbonyl)phenyl)boronic acid is a boronic acid catalyst from the Hall Group developed for chemoselective activation of oxime N−OH bonds for the preparation of functionalized amide compounds via Beckmann Rearrangement. This catalyst is efficient to transform various diaryl, aryl-alkyl, heteroaryl-alkyl, and dialkyl oximes to amide products under ambient conditions.[1]

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scope and Mechanism of a True Organocatalytic Beckmann Rearrangement with a Boronic Acid/Perfluoropinacol System under Ambient Conditions.
Mo X, et al.
Journal of the American Chemical Society, 140(15), 5264-5271 (2018)
Xiaobin Mo et al.
Journal of the American Chemical Society, 140(15), 5264-5271 (2018-03-23)
Catalytic activation of hydroxyl functionalities is of great interest for the production of pharmaceuticals and commodity chemicals. Here, 2-alkoxycarbonyl- and 2-phenoxycarbonyl-phenylboronic acid were identified as efficient catalysts for the direct and chemoselective activation of oxime N-OH bonds in the Beckmann

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service