Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

905372

Sigma-Aldrich

Pyruvic acid-1,2-13C2,d4

≥99 atom % 13C, ≥98 atom % D, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2CD4O3
Molecular Weight:
94.07
UNSPSC Code:
12142201

isotopic purity

≥99 atom % 13C
≥98 atom % D

Assay

≥99% (CP)

form

liquid

mass shift

M+6

storage temp.

−20°C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service