Skip to Content
MilliporeSigma
All Photos(1)

Documents

904589

Sigma-Aldrich

Pyruvic acid-2-13C,d4

≥99 atom % 13C, ≥97 atom % D, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC2D4O3
Molecular Weight:
93.08
UNSPSC Code:
12352106

isotopic purity

≥99 atom % 13C
≥97 atom % D

Assay

≥99% (CP)

form

liquid

mass shift

M+5

storage temp.

−20°C

SMILES string

O=C(O[2H])[13C](C([2H])([2H])[2H])=O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service