Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

904473

Sigma-Aldrich

4-Cyano-N-(trifluoromethoxy)pyridinium triflimide

≥95%

Synonym(s):

4-Cyano-1-(trifluoromethoxy)pyridin-1-ium bis((trifluoromethyl)sulfonyl)amide, Togni trifluoromethoxylation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4F9N3O5S2
CAS Number:
Molecular Weight:
469.26
UNSPSC Code:
12352101

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

4-Cyano-N-(trifluoromethoxy)pyridinium triflimide is a bench-stable trifluoromethoxylation reagent developed in the Togni lab. Under irradiation with visible-light in the presence of a photocatalyst, N-O bond cleavage results in an OCF3 radical capable of arene C-H functionalization, providing trifluoromethoxy ethers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benson J Jelier et al.
Angewandte Chemie (International ed. in English), 57(42), 13784-13789 (2018-06-22)
A simple trifluoromethoxylation method enables non-directed functionalization of C-H bonds on a range of substrates, providing access to aryl trifluoromethyl ethers. This light-driven process is distinctly different from conventional procedures and occurs through an OCF3 radical mechanism mediated by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service