Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

902802

Sigma-Aldrich

2-(2,4-Difluorophenyl)-5-fluoropyridine

≥95%

Synonym(s):

Photocatalysis ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6F3N
CAS Number:
Molecular Weight:
209.17
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

solid

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: Photocatalysis

functional group

fluoro

Application

This ligand is used in Ir(III) complexes suitable for use in photocatalysis and phosphorescent OLEDs.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service