Skip to Content
MilliporeSigma
All Photos(2)

Documents

902748

Sigma-Aldrich

Dimethyl 2-((tosyloxy)imino)malonate

≥95%

Synonym(s):

Kurti doubly N-electrophilic iminomalonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO7S
CAS Number:
Molecular Weight:
315.30
UNSPSC Code:
12352202

Assay

≥95%

form

powder or crystals

availability

available only in USA

mp

92-94 °C

Application

As demonstrated by Laszlo Kürti′s lab, ketomalonate oxime O-sulfonates act as doubly N-electrophilic linchpin reagents towards strong C-nucleophiles such as alkyl- and arylmetals (e.g., Grignard reagents). Thus, symmetrical dialkyl- as well as diarylamines may be directly prepared at low tempretatures and in the absence of transition metal catalysts. Other ketomalonate oxime O-sulfonates include 902918, 902705 and 902624.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingao Peng et al.
Organic letters, 13(19), 5244-5247 (2011-08-31)
A novel Pd(II)-catalyzed aromatic C-H ethoxycarbonylation with oxaziridine involving C-C bond cleavage is described. Various aromatic 2-phenylpyridines and related compounds as well as aryl ureas can be effectively ethoxycarbonylated. A catalytic cycle involving Pd(II) and Pd(IV) is proposed.
Padmanabha V Kattamuri et al.
Journal of the American Chemical Society, 139(32), 11184-11196 (2017-06-27)
Given the importance of amines in a large number of biologically active natural products, active pharmaceutical ingredients, agrochemicals, and functional materials, the development of efficient C-N bond-forming methods with wide substrate scope continues to be at the frontier of research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service