Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902462

Sigma-Aldrich

Molybdenum(IV) disulfide

lithium intercalated

Synonym(s):

LiMoS2, Lithium intercalated 2D-Molybdenum disulfide, Lithium intercalated moly sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$440.00

$440.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$440.00

About This Item

Empirical Formula (Hill Notation):
MoS2Li
CAS Number:
Molecular Weight:
167.01
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

$440.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

SMILES string

[Mo].[LiH].S

InChI key

CYKBBUJFMSTBEP-UHFFFAOYSA-N

Application

Owing to the thickness dependent physical and chemical properties, the transition metal dichalcogenide (TMD) based two dimensional (2D) nanomaterials have drawn significant attention recently.[1][2][3][4] The atomically thin layers of MoS2 represent important class of TMD based 2D-material. They find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service