Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902071

Sigma-Aldrich

Carboxylic acid poly(ethylene glycol)-block-poly(lactide-co-glycolide)

PEG average Mn 5000, PLGA average Mn 15000, lactide:glycolide (50:50)

Synonym(s):

Carboxylic acid-PEG-b-PLGA, Carboxylic acid-PEG-PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O3S(C2H4O)nC2H4O[(C2H2O2)x(C3H4O2)y]mH
UNSPSC Code:
12352106
NACRES:
NA.23

description

Identity: Conforms to structure (by NMR)

form

powder

feed ratio

lactide:glycolide (50:50)
lactide:glycolide:5 (50:50(by NMR))

mol wt

PEG Mn 4000-7000 g/mol (by NMR)
PEG average Mn 5000
PLGA Mn 13000-18000 g/mol (by NMR)
PLGA average Mn 15000

color

white

storage temp.

−20°C

Application

Carboxylic acid poly(ethylene glycol)-block-poly(lactide-co-glycolide) is a carboxylic acid-functionalized, biocompatible, amphiphilic block copolymer. Due to its self-assembling properties, this material can be used in the formation of nanoparticles for drug delivery. Additionally, due to the carboxylic acid-functionalization of the PEG block, this material can be used to rapidly covalently conjugate biomolecules, API, or targeting ligands. Potential uses include the targeted and/or controlled release of cancer drugs, anti-inflammatory drugs, antibiotics, or anesthetic agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradable PLGA-b-PEG polymeric nanoparticles: synthesis, properties and nanomedical applications as drug delivery system.
Locatelli E. et al
Journal of Nanoparticle Research, 14, 1316-1316 (2012)
Ilaria Monaco et al.
Journal of medicinal chemistry, 60(10), 4510-4516 (2017-05-05)
Polymeric nanoparticles (PNPs) may efficiently deliver in vivo therapeutics to tumors when conjugated to specific targeting agents. Gint4.T aptamer specifically recognizes platelet-derived growth factor receptor β and can cross the blood-brain barrier (BBB). We synthesized Gint4.T-conjugated PNPs able of high
Yihan Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1692-1716 (2016-04-22)
Poly (lactic-co-glycolic acid) (PLGA) copolymers have been broadly used in controlled drug release applications. Because these polymers are biodegradable, they provide an attractive option for drug delivery vehicles. There are a variety of material, processing, and physiological factors that impact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service