Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901889

Sigma-Aldrich

3D printing copper ink

greener alternative

Synonym(s):

3D printing copper ink, Copper, Copper ink, Cu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

viscosity

20-60 Pa.s

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Graphene based inks are ideal elements for 3D printed energy storage devices. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.
This research grade (not for human use) metallic copper 3D-printing ink can be used for printing using a direct extrusion (pneumatic or mechanical) 3D-printer into multi-layered, three-dimensional objects at room temperature. This ink should only be used in a well-ventilated environment. The extruded material rapidly solidifies and allows creation of complex, user-designed forms. Despite being mostly copper, the printed solid material is flexible (may even have limited elastic properties) and it exhibits high conductive even without post-printing curing or sintering (although the 3D-printed copper can be optionally sintered at 700 °C in non-oxidative environment to produce dense metallic 3D-printed parts). The solids loading of the as-dried material is 70 vol.% copper, 30 vol.% PLGA. This conductive copper ink can be utilized to create three-dimensional, highly conductive structures on their own, or co-3D-printed with other 3D-printing inks to create multi-material structures. The ink may also be mixed with other 3D-printing inks to compound them prior to utilization. Due to the mechanical flexibility of the material after drying, complex 2D and 3D forms can be created. This ink may also be utilized in non-3D-aprinting applications such as coating (dip-coating or direct painting), thread fabrication, or casting into molds.
Our metallic copper 3D-printing ink is comprised of copper particles (1-10 μm) suspended in a mixture of organic solvents containing a dissolved, high molecular weight elastomeric, biocompatible polyester (polylactic-co-glycolic acid; PLGA). Prior to further use, to remove residual solvents after 3D-printing (or other application such as coating, direct painting etc) the copper composite material should be washed in 70% ethanol and then by water.

Preparation Note

Prior to use mix until the ink becomes homogeneous

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service