Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901742

Sigma-Aldrich

Dimethyl[(1-pyrrolidinyl)phenyl]-amine nickel(II) chloride

≥95%

Synonym(s):

(PyrNMeNN)Ni-Cl, (SP-4-4)-Chloro[N1,N1-dimethyl-N2-[2-(1-pyrrolidinyl-κN)phenyl]-1,2-benzenediaminato-κN1N2]nickel, Second-generation Nickamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22ClN3Ni
Molecular Weight:
374.53
UNSPSC Code:
12352101

Assay

≥95%

form

powder or crystals

reaction suitability

core: nickel
reagent type: catalyst

SMILES string

CN(C)C1=C(C=CC=C1)NC2=CC=CC=C2N3CCCC3.[Ni].Cl

Application

As reported by Xile Hu′s lab, this nickel(II) pincer complex is an improvement from the first-generation Nickamine as an efficient Ni catalyst for Kumada and Suzuki-Miyaura cross-coupling of both nonactivated primary and secondary alkyl halides.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Structure-Activity Study of Nickel NNN Pincer Complexes for Alkyl-Alkyl Kumada and Suzuki-Miyaura Coupling Reactions.
Franco TD, et al.
Helvetica Chimica Acta, 99(11), 830-847 (2016)
From Dimethylamine to Pyrrolidine: The Development of an Improved Nickel Pincer Complex for Cross-Coupling of Nonactivated Secondary Alkyl Halides.
Garcia PMP, et al.
ACS Catalysis, 6 (1), 258-261 (2016)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service