Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901647

Sigma-Aldrich

Electroless nickel plating solution

ammonia type

Synonym(s):

ENPAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

form

liquid

color

blue

pH

>9

Looking for similar products? Visit Product Comparison Guide

General description

Improved standard electroless nickel plating composition specific for making ohmic contacts to silicon and other semiconductor materials.

Advantages:
  • Stable, ready to use
  • Plates uniformly on p- and n- type silicon
  • Offers excellent adhesion and solderability
  • Plates on silicon, germanium, gallium arsenide
  • Produces quality electrical contacts on p- and n- type silicon

An improved electroless nickel plating solution (Brenner type) formulated for semiconductor use. Specifically, it is designed to deposit nickel uniformly and at equal plating rates on both p- and n- type silicon. The composition of this product is based upon ions of nickel complexes and hypophosphite with stabilizers. Only high purity chemicals are used. The plating solution is a stable product ready to use without the need for additions or mixing. Transene Electroless Nickel Plating Ammonia Type operates under conditions of a catalytic oxidation-reduction reaction between nickelous and hypophosphite ions. The chemical reaction is essentially a two-step process occurring simultaneously.

H2PO2- + H2O --> H2PO3 - + 2H+ + 2e- (1)
Ni2+ + 2e- --> Ni (2)

Nickel is deposited containing about 1% phosphide which improves the physical properties of the metalization.
The plating solution has an optimized standard electrode potential of 0.44 volts. The electrode characteristics help to regulate the difference in electronegativity between p- and n- type silicon relative to the potential of the nickel complex ions. Thus the rate of deposition of nickel on p- and n- silicon is equalized. The deposition on p- and n- type silicon is 2,000 Å/minute at 90 °C.

Application

Silicon surfaces must be clean and etched in Buffer-HF or HF solution to remove oxide, rinsed and stored in alcohol. On polished silicon wafers, plate 1000-2000 A of electroless nickel; greater thickness on lapped silicon surfaces. The deposited nickel can be sintered into silicon at 500 °C to 750 °C to promote adhesion and ohmic contact. A second nickel deposit is generally required.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

Target Organs

Lungs

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service