Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901645

Sigma-Aldrich

Methoxy poly(ethylene glycol) KAT

PEG average Mn 20,000

Synonym(s):

KAT PEG 20k, mPEG KAT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CO[CH2CH2O]nCH2CH2O(C6H3BF3KNO)
UNSPSC Code:
51171641
NACRES:
NA.23

form

powder or solid

mol wt

PEG average Mn 20,000
PEG ~20,000 Da

color

off-white to pale yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Methoxy poly(ethylene glycol) KAT 20K (mPEG KAT 20K) is a monofunctional, methyl ether PEG featuring a terminal potassium acyltrifluoroborate for facile, rapid functionalization. Potassium acyltrifluoroborates (KATs) are stable functional groups that undergo rapid amide-forming ligations with hydroxylamines[1][2] in aqueous media, in the presence of unprotected functional groups. In addition to its compatibility, these reactions proceed relatively quickly[3], lending to their use with sensitive biological reagents. This conjugation reaction offers a new approach to the synthesis of complex molecules without the complication of side reactions, such protein-polymer conjugates[4]. KATs also undergo amide or imide-forming ligations in acidic conditions when reacted with primary amines or amides, respectively, as an alternative to classical acylation chemistry[5].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium Acyltrifluoroborate (KAT) Ligations are Orthogonal to Thiol-Michael and SPAAC Reactions: Covalent Dual Immobilization of Proteins onto Synthetic PEG Hydrogels.
Mazunin D, et al.
Helvetica Chimica Acta, 100(2) (2017)
Fumito Saito et al.
ACS chemical biology, 10(4), 1026-1033 (2015-01-13)
Chemoselective ligation reactions have contributed immensely to the development of organic synthesis and chemical biology. However, the ligation of stoichiometric amounts of large molecules for applications such as protein-protein conjugates is still challenging. Conjugation reactions need to be fast enough
Amide-forming ligation of acyltrifluoroborates and hydroxylamines in water.
Aaron M Dumas et al.
Angewandte Chemie (International ed. in English), 51(23), 5683-5686 (2012-04-28)
Alberto Osuna Gálvez et al.
Journal of the American Chemical Society, 139(5), 1826-1829 (2017-01-25)
Current methods for constructing amide bonds join amines and carboxylic acids by dehydrative couplings-processes that usually require organic solvents, expensive and often dangerous coupling reagents, and masking other functional groups. Here we describe an amide formation using primary amines and
Hidetoshi Noda et al.
Journal of the American Chemical Society, 136(15), 5611-5614 (2014-04-02)
The identification of fast, chemoselective bond-forming reactions is one of the major contemporary challenges in chemistry. We show that chemoselective amide-forming ligations of potassium acyltrifluoroborates (KATs) and O-carbamoylhydroxylamines proceed in the presence of all unprotected functional groups with a second-order

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service