Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901482

Sigma-Aldrich

Hexamethyldisilazane

20% solution in xylene

Synonym(s):

HMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H19NSi2
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is used to augment the adhesion of photoresist on silicon and SiO2 surfaces.
The effectiveness of HMDS on adhesion is correlated with the reactivity of this compound with surface hydroxyl groups to form a new siloxane end product, i.e. Si-O-Si(CH3)3. This newly formed termination on the substrate renders the surface more hydrophobic in character and leads to greater wettability by photoresist. The latter condition is a crucial factor in good bonding. As a result of these altered characteristics due to the surface chemistry, the treated silicon surfaces become highly compatible with both negative and positive photoresists.

Application

HMDS preparations are generally applied to the silicon wafer while spinning, prior to the application of photoresist. As an alternative procedure, the wafers may be immersed in HMDS preparations and allowed to dry after removal.

Features and Benefits

  • Ready-to-use preparations for surface treatment of silicon.
  • Promotes photoresist adhesion on silicon and SiO2 films.
  • Prevents lift-off at edges of photoresist and reduces undercutting.
  • Ensures full-line resolution.
  • Improves yields of MOS and integrated silicon devices.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

76.1 °F

Flash Point(C)

24.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service