Skip to Content
MilliporeSigma
All Photos(1)

Documents

901440

Sigma-Aldrich

Triethylphosphine solution

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

PEt3, Triethylphosphorous solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3P
CAS Number:
Molecular Weight:
118.16
Beilstein:
969170
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: ligand
reaction type: Arylations

concentration

1.0 M in 2-methyltetrahydrofuran

density

0.843 g/mL

functional group

phosphine

SMILES string

CCP(CC)CC

InChI

1S/C6H15P/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

RXJKFRMDXUJTEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Catalyst for:
  • Regioselective formation of alcohols from hydrocarbonylation of alkenes
  • Bronsted acid-assisted synthesis of dihydropyrones from aromatic aldehydes and allenoates
  • Silaboration of enynes
  • Conversion of phenylacetic acids to phenylacetonitriles of benzonitriles
  • Thermal decomposition of platinum and palladium naphthalenediyl
  • Asymmetrical Morita-Baylis-Hillman reaction
Offered as the 2-methyltetrahydrofuran solution for more convenient handling, triethylphosphine is a valuable ligand for:
  • Regioselective formation of alcohols from hydrocarbonylation of alkenes.
  • Bronsted acid-assisted synthesis of dihydropyrones from aromatic aldehydes and allenoates.
  • Silaboration of enynes.
  • Conversion of phenylacetic acids to phenylacetonitriles of benzonitriles.
  • Thermal decomposition of platinum and palladium naphthalenediyl.
  • Asymmetrical Morita-Baylis-Hillman reaction.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

8.6 °F

Flash Point(C)

-13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service