Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901209

Sigma-Aldrich

N-(But-3-yn-1-yl)-4-hydrazineylbenzamide hydrochloride

Synonym(s):

Clickable, nucleophilic hydrazine probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClN3O · xHCl
CAS Number:
Molecular Weight:
239.70 (free base basis)
UNSPSC Code:
12352101

form

powder or crystals

storage temp.

2-8°C

Application

Propylhydrazine can be used as a synthetic building blocks for various applications. Ben Cravatt′s lab recently developed clickable, nucleophilic hydrazine probes for "reverse-polarity" activity-based protein profiling, or (RP)-ABPP, where this propylhydrazine was used as a competitor for hydrazine probe 901210.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service