Skip to Content
MilliporeSigma
All Photos(2)

Documents

901207

Sigma-Aldrich

4,7-Bis(5-bromo-2-thienyl)-5,6-difluoro-2-(2-hexyldecyl)-2H-benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37Br2F2N3S2
CAS Number:
Molecular Weight:
701.57
UNSPSC Code:
12352101
NACRES:
NA.23

Assay

≥97%

form

powder or crystals

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Non-Fullerene Polymer Solar Cells Based on Alkylthio and Fluorine Substituted 2D-Conjugated Polymers Reach 9.5% Efficiency.
Bin H, et al.
Journal of the American Chemical Society, 138(13), 4657-4664 (2016)
Haijun Bin et al.
Journal of the American Chemical Society, 138(13), 4657-4664 (2016-03-22)
Non-fullerene polymer solar cells (PSCs) with solution-processable n-type organic semiconductor (n-OS) as acceptor have seen rapid progress recently owing to the synthesis of new low bandgap n-OS, such as ITIC. To further increase power conversion efficiency (PCE) of the devices

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service