Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901146

Sigma-Aldrich

TPDI-Hex

Synonym(s):

2,2′,8,8′-Tetrakis(1-ethylpropyl)-5,5′-dihexyl-[11,11′-Bi-1H-pyrrolo[2′,3′,4′,5′:4,5]phenanthro[2,1,10-def:7,8,9-def′]diisoquinoline]-1,1′,3,3′,7,7′,9,9′(2H,2′H,5H,5′H,8H,8′H)-octone, nPDI2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H80N6O8
CAS Number:
Molecular Weight:
1253.53
UNSPSC Code:
12352101

description

Band gap: 2.1 eV

form

powder

solubility

chloroform: soluble

Orbital energy

HOMO -5.8 eV 
LUMO -3.7 eV 

Legal Information

Sold under Material Transfer Agreement with Gregory Welch group at University of Calgary.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fullerene-free polymer solar cells processed from non-halogenated solvents in air with PCE of 4.8%.
Dayneko SV, et al.
Chemical Communications (Cambridge, England), 53(6), 1164-1167 (2017)
N-Annulated perylene diimide dimers: acetylene linkers as a strategy for controlling structural conformation and the impact on physical, electronic, optical and photovoltaic properties.
Cann J, et al.
Journal of Material Chemistry C, 5(8), 2074-2083 (2017)
Synthesis, Self-Assembly, and Solar Cell Performance of N-Annulated Perylene Diimide Non-Fullerene Acceptors.
Hendsbee AD, et al.
Chemistry of Materials, 28(19), 7098-7109 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service