Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901144

Sigma-Aldrich

nPDI

Synonym(s):

Hexyl N-annulated perylene diimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H41N3O4
CAS Number:
Molecular Weight:
627.77
UNSPSC Code:
12352200

description

Band gap: 2.1 eV

form

powder

solubility

chloroform: soluble

Orbital energy

HOMO -5.8 eV 
LUMO -3.7 eV 

Legal Information

Sold under Material Transfer Agreement with Gregory Welch group at University of Calgary.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey V Dayneko et al.
Chemical communications (Cambridge, England), 53(6), 1164-1167 (2017-01-06)
Progress towards practical organic solar cells amenable to large scale production is reported. Fullerene-free organic solar cells with a PCE of ∼4.8% are achieved based upon an active layer composed of the standard donor polymer PTB7-Th and a highly soluble
Synthesis, Self-Assembly, and Solar Cell Performance of N-Annulated Perylene Diimide Non-Fullerene Acceptors.
Hendsbee AD, et al.
Chemistry of Materials, 28(19), 7098-7109 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service