Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901140

Sigma-Aldrich

1,14-Diazido-3,6,9,12-tetraoxatetradecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N6O4
CAS Number:
Molecular Weight:
288.30
MDL number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry

density

1.129

storage temp.

2-8°C

SMILES string

[N-]=[N+]=NCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C10H20N6O4/c11-15-13-1-3-17-5-7-19-9-10-20-8-6-18-4-2-14-16-12/h1-10H2

InChI key

QTEOEACEOUNLRG-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service