Skip to Content
MilliporeSigma
All Photos(1)

Documents

901020

Sigma-Aldrich

Poly(dioxanone-co-glycolide)

90:10, viscosity 2.0 dL/g 

Synonym(s):

PDO-PGA, PDS-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6O3)m(C4H4O4)n
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

chips

feed ratio

p-dioxanone:glycolide 90:10±5

color

white to off-white

viscosity

1.5-2.5 dL/g
2.0 dL/g

storage temp.

−20°C

General description

Poly(dioxanone-co-glycolide) is a biodegradable copolymer synthesized from the ring-opening polymerization of p-dioxanone and glycolide. Due to the ester linkages found in the polymer backbone, these polymers are able to degrade via hydrolysis. With different degrees of aliphatic character, the ratio of the two monomers affects the rate of hydrolysis and therefore the rate of degradation. Other factors that affect the degradation of these materials could include the end application or post-polymerization processing methods.

Application

Poly(dioxanone-co-glycolide) is a biodegradable copolymer featuring the high strength of poly(dioxanone) and the flexibility of poly(glycolide) for use in biomedical research applications such as biomedical devices, drug delivery systems, and tissue engineering constructs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Main features of p-dioxanone, glycolide and lactide based copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service