Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900931

Sigma-Aldrich

3-Azidopropionic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3O2
CAS Number:
Molecular Weight:
115.09
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95%

form

liquid

reaction suitability

reaction type: click chemistry

availability

available only in USA

refractive index

n/D 1.467

solubility

DMF: soluble
DMSO: soluble
THF: soluble
chloroform: soluble
dichloromethane: soluble

density

1.2356 g/mL

storage temp.

2-8°C

InChI

1S/C3H5N3O2/c4-6-5-2-1-3(7)8/h1-2H2,(H,7,8)

InChI key

YIPVUXAMZQBALD-UHFFFAOYSA-N

Application

3-Azidopropionic acid is an azido-containing building block that can be reacted with primary amine groups in the presence of activators (e.g. EDC, or DCC), forming a stable amide bond. The terminal azide group allows conjugation with compounds containing alkyne groups through a copper(I)-catalyzed cycloaddition reaction, also known as click chemistry. 3-Azidopropionic acid is useful for crosslinking, synthesis of chemical probes, and other bioconjugation strategies.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service