Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900930

Sigma-Aldrich

6-Azidohexanoic acid

≥95%

Synonym(s):

6-Azidocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O2
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352106

Assay

≥95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

availability

available only in USA

refractive index

n/D 1.463

solubility

DMF: soluble
DMSO: soluble
THF: soluble
chloroform: soluble
dichloromethane: soluble

density

1.074 g/mL

storage temp.

2-8°C

InChI

1S/C6H11N3O2/c7-9-8-5-3-1-2-4-6(10)11/h1-5H2,(H,10,11)

InChI key

JCORXJUUSVCJEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Azidohexanoic acid is a six-carbon saturated fatty acid with an ω-terminal azide group. This building block can be reacted with primary amine groups in the presence of activators (e.g. EDC, or DCC), forming a stable amide bond. The terminal azide group allows conjugation with compounds containing alkyne groups through a copper(I)-catalyzed cycloaddition reaction, also known as click chemistry. 6-Azidohexanoic acid is useful for crosslinking, synthesis of chemical probes, and other bioconjugation strategies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated Microglia Targeting Dendrimer?Minocycline Conjugate as Therapeutics for Neuroinflammation.
Sharma R, et al.
Bioconjugate Chemistry, 28(11), 2874-2886 (2017)
Conception of Stretchable Resistive Memory Devices Based on Nanostructure?Controlled Carbohydrate?block?Polyisoprene Block Copolymers.
Hung C C, et al.
Advances in Functional Materials, 27(13) (2017)
CuAAC macrocyclization: high intramolecular selectivity through the use of copper?tris (triazole) ligand complexes.
Chouhan G and James K
Organic Letters, 13(10), 2754-2757 (2011)
Efficient Access to New Chemical Space Through Flow?Construction of Druglike Macrocycles Through Copper?Surface?Catalyzed Azide?Alkyne Cycloaddition Reactions.
Bogdan A R and James K
Chemistry?A European Journal , 16(48), 14506-14512 (2010)
Maria I Montanez et al.
Biomacromolecules, 12(6), 2114-2125 (2011-04-29)
Well-defined dendronized cellulose substrates displaying multiple representations of dual-functionality were constructed by taking advantage of the efficiency of the click reaction combined with traditional anhydride chemistry. First, activated cellulose surfaces were decorated with several generations of dendrons, and their peripheral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service