Skip to Content
MilliporeSigma
All Photos(2)

Documents

900906

Sigma-Aldrich

Solvene®300/P300

Synonym(s):

Poly(vinylidene fluoride-co-trifluoroethylene), Fluoropolymer resin, P(VDF-TrFE)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H2F2)n(C2HF3)m
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.23

description

Curie peak: 103±3 °C
MFI (melt flow index): 2-8
Piezoelectric coefficient (d33): -23 pC/N at 110Hz

form

powder

mol wt

average mol wt 300 kDa (given as an indication)

composition

TrFE, 30 mol %
VDF, 70 mol %

dielectric constant

14, 1 kHz (at 23 °C)

mp

146 °C±2 °C

InChI

1S/C2HF3.C2H2F2/c3-1-2(4)5;1-2(3)4/h1H;1H2

InChI key

XLOFNXVVMRAGLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solvene® electro-active polymer is a family of ferroelectric polymers and ferrorelaxor polymers which combines excellent electro mechanical properties with easy processing.
Solvene®300 P(VDF-TrFE) copolymers are inherently piezo-, pyro- and ferroelectric, enabling applications such as printed memories, sensors, actuators, loudspeaker, acoustic transducers, energy harvesting devices.
Solvene® materials can be processed by applying various techniques, from classic extrusion to different printing techniques on various substrates which include flexible plastic films or paper.

Legal Information

Solvene is a registered trademark of Solvay Specialty Polymers USA LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service