Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900756

Sigma-Aldrich

SLAP M

≥95%

Synonym(s):

2-((Trimethylsilyl)methoxy)ethan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17NOSi
CAS Number:
Molecular Weight:
147.29
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

storage temp.

2-8°C

SMILES string

NCCOC[Si](C)(C)C

Application

When used with aliphatic or aromatic aldehydes, this silicon amine protocol (SLAP) reagent enables the photomediated synthesis of thiomorpholines and thiazepanes. For photocatalytic cross-coupling with this SLAP Reagent, Ir[(ppy)2dtbbpy]PF6 (747769) is used with a recommended starting combination of Lewis acids Bi(OTf)3 (633305) and Cu(OTf)2 (283673) prior to subsequent substrate-specific optimization. This product was introduced in collaboration with the Bode Research Group and provides a tin-free alternative to SnAP (tin amine protocol) reagents and is well-suited for scale-up reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service