Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900715

Sigma-Aldrich

N-((S)-1-((4S,5S)-4-Benzyl-5-phenyl-4,5-dihydrooxazol-2-yl)-2,2-dimethylpropyl)acetamide

Synonym(s):

Chiral MPAO ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28N2O2
CAS Number:
Molecular Weight:
364.48
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

mp

130 °C

functional group

amide
phenyl

storage temp.

2-8°C

Application

N-((S)-1-((4S,5S)-4-Benzyl-5-phenyl-4,5-dihydrooxazol-2-yl)-2,2-dimethylpropyl)acetamide is a chiral ligand developed by Jin-Quan Yu′s lab for Pd(II)-catalyzed enantioselective β-C(sp3)H arylation, alkenylation, and alkynylation. This mono-protected aminomethyl oxazoline (MPAO) ligand was used in the challenging desymmetrization of isopropyl groups.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service