Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900553

Sigma-Aldrich

Potassium hex-5-enoyltrifluoroborate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9BF3KO
CAS Number:
Molecular Weight:
204.04
UNSPSC Code:
12352103

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: ligand

functional group

allyl

Application

Potassium acyltrifluroborates (KATs) are bench-, air-, and moisture-stable reagents for rapid chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service