Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900539

Sigma-Aldrich

Ir[p-F(t-Bu)-ppy]3

Synonym(s):

tris[2-[4-(1,1-dimethylethyl)-2-pyridinyl-κN]-5-fluorophenyl-κC]- iridium

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$196.00

$196.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$196.00

About This Item

Empirical Formula (Hill Notation):
C45H45F3IrN3
CAS Number:
Molecular Weight:
877.07
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$196.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder or crystals

Quality Level

reaction suitability

core: iridium
reaction type: Photocatalysis
reagent type: catalyst

mp

372.5 °C

photocatalyst activation

430 nm

SMILES string

[Ir](c5c(ccc(c5)F)c6nccc(c6)C(C)(C)C)(c3c(ccc(c3)F)c4nccc(c4)C(C)(C)C)c1c(ccc(c1)F)c2nccc(c2)C(C)(C)C

InChI

1S/3C15H15FN.Ir/c3*1-15(2,3)12-8-9-17-14(10-12)11-4-6-13(16)7-5-11;/h3*4,6-10H,1-3H3;

Application

Ir[p-F(t-Bu)-ppy]3 is a photocatalyst which readily facilitates the decarboxylative arylation of α amino acids using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service