Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900339

Sigma-Aldrich

Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-maleimide

lactide:glycolide 50:50, PEG average Mn 5000, PLGA average Mn 20,000

Synonym(s):

PLGA-PEG-MAL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H11N2O3(C2H4O)x(C3H4O2)y(C2H2O2)zH
UNSPSC Code:
12352200
NACRES:
NA.23

form

chunks

Quality Level

mol wt

PEG average Mn 5000
PLGA average Mn 20,000

composition

lactide:glycolide 50:50

storage temp.

−20°C

Application

Diblock polymer can form micelles by emulsion techniques or microfluidic methods. The maleimide functional group may be used for conjugation to specific binding ligands via thiol-maleimide reaction.

Other Notes

Chapter 5: “Nanobody-Targeted and RNase-Loaded Nanoparticles Based on a Hydrophilic Polyester Aimed for Cancer Therapy” in Samadi, N.
Biodegradable Nanoparticles Based on Aliphatic Polyesters, Towards Targeted Intracellular Delivery of Protein Therapeutics. Ph.D. Thesis, Utrecht University, Utrecht, Netherlands. 2014.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LyP-1-conjugated nanoparticles for targeting drug delivery to lymphatic metastatic tumors.
Luo G, et al.
International Journal of Pharmaceutics, 385(1-2), 150-156 (2010)
Drug release from PLA/PEG microparticulates.
Huang YY, et al.
International Journal of Pharmaceutics, 156(1), 9-15 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service