Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900332

Sigma-Aldrich

CdS/ZnS core-shell type quantum dots

lyophilized, fluorescence λem 450 nm, solid

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

lyophilized
powder
solid

λmax

435 nm

fluorescence

λem 450 nm
λem 450 nm±10 nm, quantum yield >50%

functional group

carboxylic acid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

CdS/ZnS or CdSe/ZnS quantum dots (QDs) with carboxylic acid in lyophilized solid form can readily be reconstituted in DI water or DMSO to any desired concentration and are very stable in most buffer solutions in the pH range of 5-10. These QDs possess zeta potential in -30 mV to -50 mV range and can easily be reacted with amine containing molecules in DMSO or water. These QDs find applications in bioimaging and biosensing.[1][2][3]

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Xu et al.
ACS applied materials & interfaces, 5(8), 2901-2907 (2013-04-05)
The excellent optical properties of quantum dots (QDs), such as high brightness, high photostability, continuous absorption, and narrow emission bandwidth, make them ideal as optical labels to develop QD-based immunohistofluorescence (IHF) imaging for multiplexing cancer biomarker detection on formalin-fixed and
Zengyan Wei et al.
Soft matter, 8(26), 6871-6875 (2012-09-18)
Genetically-engineered collagen peptides were assembled into freestanding films when QDs are co-assembled as joints between collagen domains. These peptide based films show excellent mechanical properties with Young's modulus of ~20 GPa, much larger than most of multi-composite polymer films and
Lily Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 5(2), 235-243 (2008-12-18)
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles

Articles

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service