Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900167

Sigma-Aldrich

UV curable resin

rapid cure

Synonym(s):

CPS 1040 UV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

viscosity

475 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)

density

1.24 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35 °C.
  • Keep tightly closed. Store in a cool, dry, well-ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80 °C), pressure, and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.

Application

  • Cures extremely rapidly to form a glassy, high modulus material
  • Excellent for use in optical applications
  • High refractive index - 1.54
  • High impact absorption
  • Non-yellowing
  • No oxygen inhibition.
  • Mechanical properties after curing:
Modulus*: 930 MPa
Strength*: 34 MPa
Elongation*: 13%
Pencil Hardness: 6H
Tack Free**: <2 min

*Test Resources: tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service