Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900164

Sigma-Aldrich

UV curable resin

rapid cure, adhesible to plastics and glass

Synonym(s):

CPS 1040 UV-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

liquid (colorless)

viscosity

341 cP (Brookfield, size 31 spindle)

transition temp

flash point 110 °C

density

1.17 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35 °C.
  • Keep tightly closed. Store in a cool, dry, well-ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80 °C), pressure and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.

Application

  • Cures extremely rapidly to form a glassy, high modulus material.
  • Excellent for use in optical applications.
  • High refractive index - 1.54.
  • High impact absorption.
  • Non-yellowing.
  • No oxygen inhibition.
  • Mechanical properties after curing:
Modulus*: 150MPa
Strength*: 13MPa
Elongation*: 61%
Tack Free**: 130 fpm

*Test resources: Tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service