Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900157

Sigma-Aldrich

2-Cyanobutanyl-2-yl 3,5-dimethyl-1H-pyrazole-1-carbodithioate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N3S2
Molecular Weight:
253.39
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

storage condition

protect from light

mp

117-122 °C

SMILES string

S=C(SC(CC)(C)C#N)N1N=C(C)C=C1C

InChI

1S/C11H15N3S2/c1-5-11(4,7-12)16-10(15)14-9(3)6-8(2)13-14/h6H,5H2,1-4H3

InChI key

YLQFEMAFHJDHDH-UHFFFAOYSA-N

Application

A stable, dithiocarbamate chain transfer agent (CTA), also referred to as a RAFT agent, that can control chain growth in free radical polymerization, producing polymers with well-defined molecular weights, and low polydispersities. 2-Cyanobutanyl-2-yl 3,5-dimethyl-1H-pyrazole-1-carbodithioate can be used to control homo-polymerisation and co-polymerisation of acrylic acid, acrylates, acrylamides, and styrenes (20 MAMs), and LAMs such as vinyl acetate. Additionally, it can be used to polymerise methacrylates and methacrylamides, with good MW control, and broad polydispersities. It can be used to make MAM-LAM block copolymers. It is soluble in organic media.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478, WO99/311444.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dithiocarbamate RAFT agents with broad applicability ? the 3,5-dimethyl-1H-pyrazole-1-carbodithioates.
Polym. Chem., 7, 481-492 (2016)

Articles

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service