Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900087

Sigma-Aldrich

2-(Ethyl-2,2,2-d3) toluene

≥97 atom % D, ≥98% (CP)

Synonym(s):

1-(Ethyl-2,2,2-d3)-4-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D3H9
Molecular Weight:
123.21
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

≥97 atom % D

Assay

≥98% (CP)

form

liquid

mass shift

M+3

storage temp.

2-8°C

SMILES string

CC1=C(CC([2H])([2H])[2H])C=CC=C1

InChI

1S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7H,3H2,1-2H3/i1D3

InChI key

HYFLWBNQFMXCPA-FIBGUPNXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service