Skip to Content
MilliporeSigma
All Photos(1)

Documents

900065

Sigma-Aldrich

(±)-1,3-Butanediol-1,3-13C2

≥99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C2H10O2
Molecular Weight:
92.11
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

liquid

mass shift

M+2

InChI

1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3/i3+1,4+1

InChI key

PUPZLCDOIYMWBV-CQDYUVAPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service