Skip to Content
MilliporeSigma
All Photos(2)

Documents

900016

Sigma-Aldrich

TBAF(9-Ph-9-fluorenol)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H78FNO3
Molecular Weight:
1036.40
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

138-143 °C

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.OC1(C2=CC=CC=C2)C3=C(C=CC=C3)C4=C1C=CC=C4.OC5(C6=CC=CC=C6)C7=C(C=CC=C7)C8=C5C=CC=C8.OC9(C%10=CC=CC=C%10)C%11=C(C=CC=C%11)C%12=C9C=CC=C%12.[F-]

InChI

1S/3C19H14O.C16H36N.FH/c3*20-19(14-8-2-1-3-9-14)17-12-6-4-10-15(17)16-11-5-7-13-18(16)19;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h3*1-13,20H;5-16H2,1-4H3;1H/q;;;+1;/p-1

InChI key

HTKORNSKGASQCX-UHFFFAOYSA-M

General description

TBAF(9-Ph-9-fluorenol)3 is a fluoride–alcohol complex. It is formed by reacting 9-phenylfluoren-9-ol with tetrabutylammonium fluoride trihydrate (TBAF(H2O)3). The geometric structure and bonding of TBAF(9-Ph-9-fluorenol)3 have been investigated.

Application

Nucleophilic fluorinating reagent is a more convenient crystalline solid for handling and storage compared to tetrabutylammonium fluoride hydrate (Aldrich 241512). As reported by Engle and Gouverneur, these solids display comparable reactivity on similar substrates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coordination diversity in hydrogen-bonded homoleptic fluoride?alcohol complexes modulates reactivity.
Engle KM, et al.
Chemical Science, 6(9), 5293-5302 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service