Skip to Content
MilliporeSigma
All Photos(1)

Documents

900015

Sigma-Aldrich

Trimethylpentafluoroethylsilane

97%

Synonym(s):

(Pentafluoroethyl)trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9F5Si
CAS Number:
Molecular Weight:
192.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n/D 1.325

density

1.095 g/mL

SMILES string

C[Si](C)(C)C(F)(F)C(F)(F)F

InChI

1S/C5H9F5Si/c1-11(2,3)5(9,10)4(6,7)8/h1-3H3

InChI key

MTPVUVINMAGMJL-UHFFFAOYSA-N

General description

Trimethylpentafluoroethylsilane ((Pentafluoroethyl)trimethylsilane) is a perfluoroalkylsilane. Alkyl triflates undergo nucleophilic pentafluoroethylation with trimethylpentafluoroethylsilane to form the corresponding pentafluoroethylated alkanes.

Application

Trimethylpentafluoroethylsilane ((Pentafluoroethyl)trimethylsilane) may be used as a perfluoroalkylating reagent for the synthesis of the following quinoline derivatives:
  • 5-bromo-2-(perfluoroethyl)quinoline
  • 8-methoxy-2-(perfluoroethyl)quinoline
  • 1-(perfluoroethyl)isoquinoline
  • 8-(tert-butoxy)-5,7-dichloro-2-(perfluoroethyl)quinolone
It may also be used in the synthesis of pentafluoroethylcyclohexanes.
Trimethylpentafluoroethylsilane has been reported as a Ruppert-Prakash type reagent for the addition of pentafluoroethane. Recent report by Larionov and coworkers displayed the feasibility of adding pentrafluoroethane to N-heterocycles under basic conditions.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup - (calculated)

Flash Point(C)

-17 °C - closed cup - (calculated)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile New Method for the Two-step Substitution of Hydroxy Groups in Primary Alcohols for Trifluoromethyl and Pentafluoroethyl Moieties.
Sevenard DV, et al.
Synlett, 2001(03), 0379-0381 (2001)
Experimental determination of the conformational free energies (A values) of fluorinated substituents in cyclohexane by dynamic 19 F NMR spectroscopy. Part 2. Extension to fluoromethyl, difluoromethyl, pentafluoroethyl, trifluoromethylthio and trifluoromethoxy groups.
Carcenac Y, et al.
New. J. Chem., 30(3), 447-457 (2006)
David E Stephens et al.
Organic & biomolecular chemistry, 12(32), 6190-6199 (2014-07-06)
The scope and mechanistic implications of the direct transformation of heterocyclic N-oxides to 2-trifluoromethyl-, and related perfluoroalkyl- and perfluoroaryl-substituted N-heterocycles has been studied. The reaction is effected by perfluoroalkyl- and perfluorophenyltrimethylsilane in the presence of strong base. In situ displacement

Related Content

The major research interests of Prof. Jinbo Hu's lab include the development of new fluorination reagents and reactions, especially the difluoromethylation, difluoromethylenation, and monofluoromethylation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service