Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

89799

Sigma-Aldrich

1,3-Di-Boc-2-(2-hydroxyethyl)guanidine

≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H25N3O5
CAS Number:
Molecular Weight:
303.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (HPLC)

form

powder

mp

120-125 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N\C(NC(=O)OC(C)(C)C)=N\CCO

InChI

1S/C13H25N3O5/c1-12(2,3)20-10(18)15-9(14-7-8-17)16-11(19)21-13(4,5)6/h17H,7-8H2,1-6H3,(H2,14,15,16,18,19)

InChI key

NHRGPOQWQFSEOH-UHFFFAOYSA-N

Other Notes

Guanidinylation reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Feichtinger et al.
The Journal of Organic Chemistry, 63, 8432-8432 (1998)
K C Nicolaou et al.
Bioorganic & medicinal chemistry, 6(8), 1185-1208 (1998-10-24)
Recent studies demonstrated that peptide and antibody antagonists of integrin alpha v beta 3 block angiogenesis and tumor growth. In this article, the design, synthesis and biological evaluation of a series of nitroaryl ether-based, nonpeptide mimetics are described. The design
Antoine Frère et al.
Biomacromolecules, 16(3), 769-779 (2015-01-21)
RNAi therapeutics are promising therapeutic tools that have sparked the interest of many researchers. In an effort to provide a safe alternative to PEI, we have designed a series of new guanidinium- and morpholino-functionalized biocompatible and biodegradable polycarbonate vectors. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service