Skip to Content
MilliporeSigma
All Photos(1)

Documents

89235

Sigma-Aldrich

(−)-Thujopsene

≥97.0% (GC)

Synonym(s):

(1S,6S,10S)-2,2,6,9-Tetramethyltricyclo[8.1.0.01.6]undec-8-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
1907465
EC Number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

refractive index

n20/D 1.505

bp

258-260 °C (lit.)

density

0.936 g/mL at 20 °C (lit.)

SMILES string

CC1=CC[C@]2(C)CCCC(C)(C)[C@@]23C[C@@H]13

InChI

1S/C15H24/c1-11-6-9-14(4)8-5-7-13(2,3)15(14)10-12(11)15/h6,12H,5,7-10H2,1-4H3/t12-,14-,15-/m0/s1

InChI key

WXQGPFZDVCRBME-QEJZJMRPSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franck A Ditengou et al.
Nature communications, 6, 6279-6279 (2015-02-24)
The mutualistic association of roots with ectomycorrhizal fungi promotes plant health and is a hallmark of boreal and temperate forests worldwide. In the pre-colonization phase, before direct contact, lateral root (LR) production is massively stimulated, yet little is known about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service