Skip to Content
MilliporeSigma
All Photos(1)

Documents

89048

Sigma-Aldrich

Fmoc-Arg(Pbf)-OPfp

technical, ≥80% (HPLC)

Synonym(s):

N-α-Fmoc-NG-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H39N4F5O7S
CAS Number:
Molecular Weight:
814.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

grade

technical

Assay

≥80% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

[H][C@@](CCCNC(=N)NS(=O)(=O)c1c(C)c(C)c2OC(C)(C)Cc2c1C)(NC(=O)OCC3c4ccccc4-c5ccccc35)C(=O)Oc6c(F)c(F)c(F)c(F)c6F

InChI

1S/C40H39F5N4O7S/c1-19-20(2)36(21(3)26-17-40(4,5)56-34(19)26)57(52,53)49-38(46)47-16-10-15-28(37(50)55-35-32(44)30(42)29(41)31(43)33(35)45)48-39(51)54-18-27-24-13-8-6-11-22(24)23-12-7-9-14-25(23)27/h6-9,11-14,27-28H,10,15-18H2,1-5H3,(H,48,51)(H3,46,47,49)/t28-/m0/s1

InChI key

SKUZYOYASRPHMO-NDEPHWFRSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service