Skip to Content
MilliporeSigma
All Photos(1)

Documents

88754

Sigma-Aldrich

(S)-1-[(RP)-2-(Di-tert-butylphosphino)ferrocenyl]ethyldiphenylphosphine

≥97%

Synonym(s):

(1S)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J502-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40FeP2
CAS Number:
Molecular Weight:
542.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(C(C)(C)C)C(C)(C)C)P(c3ccccc3)c4ccccc4

InChI

1S/C27H35P2.C5H5.Fe/c1-21(24-19-14-20-25(24)29(26(2,3)4)27(5,6)7)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;1-2-4-5-3-1;/h8-21H,1-7H3;1-5H;/t21-;;/m0../s1

InChI key

MJOXBJGLXGTJRJ-FGJQBABTSA-N

Related Categories

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Research in the Togni group focuses on the development of new ligands and reagents. These two general directions thus impact the ability to construct molecules in more efficient or unprecedented ways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service