88738
(S)-1-[(RP)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}ethyldi-tert-butylphosphine
≥97%
Synonym(s):
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-[bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocene (acc to CAS), Josiphos SL-J013-2
About This Item
Assay
≥97%
form
powder
optical purity
ee: ≥99%
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.COc2c(C)cc(cc2C)P([C]3[CH][CH][CH][C]3[C@H](C)P(C(C)(C)C)C(C)(C)C)c4cc(C)c(OC)c(C)c4
InChI
1S/C33H47O2P2.C5H5.Fe/c1-21-17-26(18-22(2)30(21)34-12)36(27-19-23(3)31(35-13)24(4)20-27)29-16-14-15-28(29)25(5)37(32(6,7)8)33(9,10)11;1-2-4-5-3-1;/h14-20,25H,1-13H3;1-5H;/t25-;;/m0../s1
InChI key
UALLLNAXGQTBJA-WLOLSGMKSA-N
General description
Packaging
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service