Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88724

Sigma-Aldrich

(S)-1-[(Rp)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

≥97%

Synonym(s):

(1S)-1-(Dicyclohexylphosphino)-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J004-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44FeP2
CAS Number:
Molecular Weight:
594.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97%

form

crystals

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(C3CCCCC3)C4CCCCC4)P(c5ccccc5)c6ccccc6

InChI

1S/C31H39P2.C5H5.Fe/c1-25(32(26-15-6-2-7-16-26)27-17-8-3-9-18-27)30-23-14-24-31(30)33(28-19-10-4-11-20-28)29-21-12-5-13-22-29;1-2-4-5-3-1;/h2-3,6-9,14-18,23-25,28-29H,4-5,10-13,19-22H2,1H3;1-5H;/t25-;;/m0../s1

InChI key

AWOOIDCKLIQAJV-WLOLSGMKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service