Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88298

Sigma-Aldrich

(±)-Theaspirane

technical, ≥90% (GC)

Synonym(s):

2,6,10,10-Tetramethyl-1-oxaspiro[4.5]dec-6-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O
CAS Number:
Molecular Weight:
194.31
Beilstein:
1424383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.438 (lit.)
n20/D 1.488

bp

68-72 °C/3 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CC1CCC2(O1)C(C)=CCCC2(C)C

InChI

1S/C13H22O/c1-10-6-5-8-12(3,4)13(10)9-7-11(2)14-13/h6,11H,5,7-9H2,1-4H3

InChI key

GYUZHTWCNKINPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service