Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88074

Sigma-Aldrich

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate

≥98.0% (TLC)

Synonym(s):

meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulfonic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H26N4Na4O12S4 · xH2O
CAS Number:
Molecular Weight:
1022.92 (anhydrous basis)
Beilstein:
5488862
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

crystals

reaction suitability

core: porphyrin
reaction type: Photocatalysis
reagent type: catalyst

impurities

water (report result)

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(cc4)S([O-])(=O)=O)c5ccc([nH]5)c(-c6ccc(cc6)S([O-])(=O)=O)c7ccc(n7)c(-c8ccc(cc8)S([O-])(=O)=O)c9ccc2[nH]9

InChI

1S/C44H30N4O12S4.4Na.H2O/c49-61(50,51)29-9-1-25(2-10-29)41-33-17-19-35(45-33)42(26-3-11-30(12-4-26)62(52,53)54)37-21-23-39(47-37)44(28-7-15-32(16-8-28)64(58,59)60)40-24-22-38(48-40)43(36-20-18-34(41)46-36)27-5-13-31(14-6-27)63(55,56)57;;;;;/h1-24,45,48H,(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60);;;;;1H2/q;4*+1;/p-4/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;;;;

InChI key

LMNVUEGSCDLLCZ-FLYAKZGESA-J

Looking for similar products? Visit Product Comparison Guide

General description

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

max. Absorption (in Methanol): 412-416 nm

Other Notes

Metal complexes are employed in very sensitive chemiluminescence analyses

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Motsenbocker et al.
Analytical Chemistry, 65, 397-397 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service