Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87743

Sigma-Aldrich

Tetramethylammonium sulfate hydrate

Synonym(s):

Bis(tetramethylammonium) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)4N]2(SO4) · xH2O
CAS Number:
Molecular Weight:
244.35 (anhydrous basis)
Beilstein:
3918039
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

SMILES string

O.C[N+](C)(C)C.C[N+](C)(C)C.[O-]S([O-])(=O)=O

InChI

1S/2C4H12N.H2O4S.H2O/c3*1-5(2,3)4;/h2*1-4H3;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

SMBVAMYBVVNSBZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Chen et al.
Bioresource technology, 117, 186-192 (2012-05-23)
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS)
Yasuyuki Ishida et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(10), 1053-1056 (2011-10-12)
Thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate was applied to the direct determination of terephthalic acid (TPA) contained in solid decomposition products obtained from the hydrothermal recycling process of poly(ethylene terephthalate) (PET). On the chromatograms of the hydrothermal decomposition
Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Marta Matvienko et al.
PloS one, 8(2), e55913-e55913 (2013-02-15)
Several applications of high throughput genome and transcriptome sequencing would benefit from a reduction of the high-copy-number sequences in the libraries being sequenced and analyzed, particularly when applied to species with large genomes. We adapted and analyzed the consequences of
Federica Maraschi et al.
Analytical biochemistry, 426(2), 142-146 (2012-05-01)
This article presents the first results demonstrating that total silicon trace concentration in human ventricular whole blood may be used as a further marker in the diagnosis of drowning. The difference in silicon content between the left and right ventricles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service