Skip to Content
MilliporeSigma
All Photos(1)

Documents

87652

Sigma-Aldrich

Tetrakis[tris(dimethylamino)phosphoranylidenamino]phosphonium fluoride solution

~0.3 M in benzene

Synonym(s):

Phosphazenium fluoride P5-F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H72FN16P5
CAS Number:
Molecular Weight:
758.80
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

form

solution

concentration

~0.3 M in benzene

SMILES string

[F-].CN(C)P(=N[P+](N=P(N(C)C)(N(C)C)N(C)C)(N=P(N(C)C)(N(C)C)N(C)C)N=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C24H72N16P5.FH/c1-29(2)42(30(3)4,31(5)6)25-41(26-43(32(7)8,33(9)10)34(11)12,27-44(35(13)14,36(15)16)37(17)18)28-45(38(19)20,39(21)22)40(23)24;/h1-24H3;1H/q+1;/p-1

InChI key

YZLHWBJVVXMBGF-UHFFFAOYSA-M

Other Notes

Extremely strong and lipophilic "naked" fluoride source which can be dried without decomposition

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Skin Corr. 1B - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Schwesinger et al.
GIT, 897-897 (1992)
M. Wollenweber et al
Angewandte Chemie (International Edition in English), 106, 84-84 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service