Skip to Content
MilliporeSigma
All Photos(1)

Documents

87477

Sigma-Aldrich

2-(Tetrahydro-2H-pyran-2-yloxy)ethanol

≥98.0% (GC)

Synonym(s):

2-(2-Hydroxyethoxy)tetrahydropyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
Beilstein:
1280440
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.457

bp

95 °C/22 mmHg (lit.)

density

1.077 g/mL at 20 °C (lit.)

SMILES string

OCCOC1CCCCO1

InChI

1S/C7H14O3/c8-4-6-10-7-3-1-2-5-9-7/h7-8H,1-6H2

InChI key

XDBZJXRPEKFIFR-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service