Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87197

Sigma-Aldrich

1-Tetradecyne

≥97.0%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$225.00

$225.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$225.00

About This Item

Linear Formula:
CH3(CH2)11C≡CH
CAS Number:
Molecular Weight:
194.36
Beilstein:
1746546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$225.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.440

density

0.790 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCC#C

InChI

1S/C14H26/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h1H,4-14H2,2H3

InChI key

DZEFNRWGWQDGTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janneke Veerbeek et al.
ACS applied materials & interfaces, 9(1), 413-421 (2016-12-10)
Silicon-based solar fuel devices require passivation for optimal performance yet at the same time need functionalization with (photo)catalysts for efficient solar fuel production. Here, we use molecular monolayers to enable electrical passivation and simultaneous functionalization of silicon-based solar cells. Organic
Timotej Strmeň et al.
Rapid communications in mass spectrometry : RCM, 34(10), e8722-e8722 (2020-01-09)
Hyphenation of atmospheric pressure chemical ionization (APCI) mass spectrometry with capillary and micro high-performance liquid chromatography (HPLC) is attractive for many applications, but reliable ion sources dedicated to these conditions are still missing. There are a number of aspects to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service