Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86918

Sigma-Aldrich

Tetrabutylammonium triiodide

≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(I3)
CAS Number:
Molecular Weight:
623.18
Beilstein:
3743192
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (AT)

form

crystals

mp

69-71 °C

SMILES string

I[I-]I.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.I3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-3-2/h5-16H2,1-4H3;/q+1;-1

InChI key

SBSSZSCMFDYICE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Abe et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 212, 255-261 (2019-01-18)
In this study, the effect of alcohol on polyiodide formation in room-temperature ionic liquid (RTIL) was examined by time evolutions of Raman spectra in the low-frequency region and by color changes of the sample. The RTIL was 1-methyl-3-propylimidazolium iodide, [C3mim][I].
Rui Xian et al.
Nature chemistry, 9(6), 516-522 (2017-05-26)
Solid-state reactions are influenced by the spatial arrangement of the reactants and the electrostatic environment of the lattice, which may enable lattice-directed chemical dynamics. Unlike the caging imposed by an inert matrix, an active lattice participates in the reaction, however
Yi-June Huang et al.
Nanoscale, 11(26), 12507-12516 (2019-05-30)
In this work, pristine carbon aerogels (CAs) were used as Pt-free counter electrodes (CEs) in dye-sensitized solar cells (DSSCs) by varying the molar ratio of their precursors. Pristine mesoporous CAs with controlled resorcinol (R)/formaldehyde (F) and resorcinol (R)/sodium carbonate (C)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service